missing translation for 'onlineSavingsMsg'
Learn More

Heptane, CHROMASOLV™, ≥99%, LC-MS, Honeywell Riedel-de Haën™

Product Code. 15634770
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Density 0.684g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Linear Formula CH3(CH2)5CH3
Vapor Pressure 83 mmHg (37.7°C), 40 mmHg (20°C)
UN Number UN1206
Formula Weight 100.2
Percent Purity ≥99%
Chemical Name or Material Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'