missing translation for 'onlineSavingsMsg'
Learn More

Hexane, ReagentPlus™, ≥99%, Honeywell™

Reagent Grade,≥99%

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15661380
View Documents Promotion Details
Honeywell Research Chemicals
139386-100ML
100 mL Glass Bottle N/A
N/A
15681380
View Documents Promotion Details
Honeywell Research Chemicals
139386-500ML
500 mL Glass Bottle N/A
N/A
15671380
View Documents Promotion Details
Honeywell Research Chemicals
139386-2.5L
2.5 L Glass Bottle N/A
N/A
15691380
View Documents Promotion Details
Honeywell Research Chemicals
139386-6X500ML
6 x 500 mL Glass Bottle N/A
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-95°C
Colorless
66°C to 69°C
−26°C (−14.8°F)
Liquid
C6H14
MFCD02179311
UN1208
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18g/mol
ReagentPlus™
110-54-3
0.659g/mL at 25°C
0.077
453°F
n20/D 1.375(lit.)
CH3(CH2)4CH3
5.2psi (37.7°C), 132mmHg (20°C), 256mmHg (37.7°C)
1730733
Insoluble in water
CCCCCC
86.18
CHEBI:29021
≥99%
Hexane
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P370 + P378-P403 + P235

H225-H304-H315-H336-H361f-H373-H411

missing translation for 'dotInformation' : class 3 - PG 2 - Hexanes

missing translation for 'einecsNumber' : 203-777-6

missing translation for 'rtecsNumber' : MN9275000

Product Identifier
  • Hexane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : n-hexane

SDS
Documents

Documents

Product Certifications