missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, ≈95% N-Hexane, for HPLC
174.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
Description
C6H14, CAS Number-110-54-3, hexanen, hexan, heksan, esani, dipropyl, skellysolve b, hexyl hydride, n-hexane, gettysolve-b, hexanes, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 69°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 160mbar at 20°C |
Viscosity | 0.31 mPaS at 20°C |
Formula Weight | 86.18g/mol |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Hexanes, 95% n-Hexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Reproductive toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H361f-Suspected of damaging fertility.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'