missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ≈95% N-Hexane, HPLC for Fluorescence Detection

82.30 € - 199.00 €

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009520
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10647602
View Documents
Fisher Chemical
H/0409/PB15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10409370
View Documents
Fisher Chemical
H/0409/PB17
2.5 L
199.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexanes, ≈95% N-Hexane, HPLC for Fluorescence Detection

Specifications

Chemical Identifiers

92112-69-1
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD00009520
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-95°C
Colorless
Plastic-coated Glass Bottle
C6H14
160mbar at 20°C
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.18
CHEBI:29021
92112-69-1
69°C
Liquid
MFCD00009520
0.31 mPaS at 20°C
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hexane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications