Learn More
Hexanes, 98+%, extra pure, mixture of isomers, Thermo Scientific Chemicals
49.65 € - 512.55 €
Specifications
Chemical Name or Material | Hexanes |
---|---|
Melting Point | -95.0°C |
Name Note | Mixture of Isomers |
CAS | 92112-69-1, 110-54-3 |
Color | Colorless to Yellow |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10572391
|
Thermo Scientific Acros
176660010 |
1 L | Glass Bottle |
49.65 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409650
|
Thermo Scientific Acros
176660025 |
2.5 L | Glass Bottle |
92.86 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10773884
|
Thermo Scientific Acros
176660250 |
25 L | Metal Drum |
512.55 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanes | |
Mixture of Isomers | |
Colorless to Yellow | |
69.0°C | |
Liquid or Solid | |
98+% | |
0.659 | |
CH3(CH2)4CH3 | |
0.31 mPa.s (20°C) | |
0.05% Max. | |
Hex | |
86.18 | |
Extra Pure |
-95.0°C | |
92112-69-1, 110-54-3 | |
0.6590g/mL | |
−22°C | |
1.3748 to 1.3810 | |
0.001 % max. | |
C6H14 | |
MFCD00009520 | |
01, 142 | |
14, 4694 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
≥98% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.
Danger
- Hexanes
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Hexane, branched andlinear
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.