Learn More
Hexanes, 99%, for spectroscopy ACS, mixture of isomers, Thermo Scientific Chemicals
47.20 € - 159.00 €
Specifications
Chemical Name or Material | Hexanes |
---|---|
Melting Point | -95.0°C |
Name Note | Mixture of isomers, for spectroscopy |
Absorbance | 0.01 max. at 280nm, 0.01 max. at 400nm, 0.02 max. at 250nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 0.3 max. at 210nm, 1 max. at 195nm |
CAS | 92112-69-1, 110-54-3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10371201
|
Thermo Scientific Acros
411555000 |
500 mL | EcoSafPak™ Glass Bottle |
47.20 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10127333
|
Thermo Scientific Acros
411550025 |
2.5 L | Glass Bottle |
159.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
Hexanes | |
Mixture of isomers, for spectroscopy | |
92112-69-1, 110-54-3 | |
69.0°C | |
Liquid | |
99% | |
0.659 | |
0.0003 meq/g max. | |
MFCD00009520 | |
0.31 mPa.s (20°C) | |
14, 4694 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
99% |
-95.0°C | |
0.01 max. at 280nm, 0.01 max. at 400nm, 0.02 max. at 250nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 0.3 max. at 210nm, 1 max. at 195nm | |
0.6590g/mL | |
−22°C | |
1.3748 to 1.3810 | |
0.001% max. | |
C6H14 | |
CH3(CH2)4CH3 | |
Vapor Pressure: 160mbar at 20°C | |
01, 142 | |
Hex | |
86.18 |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 203-777-6
missing translation for 'rtecsNumber' : MN9275000
missing translation for 'tsca' : TSCA
- Hexanes
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only