missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Description
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 69°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 160mbar at 20°C |
Viscosity | 0.31 mPaS at 20°C |
Formula Weight | 86.18g/mol |
Documents
Scientific Resources
Brochures
Cosmetic and chemical industries
File size: 3.8 MB
Safety and Handling




Product Identifier
- Hexanes, approx. 95% n-hexane
- Danger
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H361f-Suspected of damaging fertility.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Hexane
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction