missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, HPLC Grade, 99+%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 045652.K7

 View more versions of this product

Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15452427

  • 186.00 € / 4 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Hexanes, mixed isomers are used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hexanes
92112-69-1, 110-54-3, 96-14-0, 107-83-5, 96-37-7, 71-43-2
Colorless liquid
65°C to 70°C
<-23°C (-9°F)
Liquid
>99%
MFCD02179311
Miscible with ether,chloroform,alcohol,and acetone. Immiscible with water.
HPLC
-95°C
Colorless
0.67 g/cm3
4 L
240°C (464°F)
1.3795
C6H14
160 hPa (120mm Hg) at 20°C
86.18
n-Hexane: 65.0%; 3-Methylpentane: 25.0%; 2-Methylpentane: 5.0%; Methylcyclopentane: 4.999%; Benzene: 0.001%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only