missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 210590250

217.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10125263

  • 218.09 € / 25 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hexanes
Mixture of isomers
0.6590g/mL
25 L
Metal Drum
1.3748 to 1.3810
0.659
CH3(CH2)4CH3
0.31 mPa.s (20°C)
14, 4694
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Technical
-95.0°C
92112-69-1, 110-54-3
69.0°C
−22°C
Liquid
0.002% max.
C6H14
MFCD00009520
01, 142
Hex
86.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hexanes

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only