missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic acid, 99%

Product Code. 10626204 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10626204

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-62-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004421
InChI Key FUZZWVXGSFPDMH-UHFFFAOYSA-N
Synonym caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid
PubChem CID 8892
ChEBI CHEBI:30776
IUPAC Name hexanoic acid
SMILES CCCCCC(=O)O

Specifications

Acidity 473 to 493mg KOH/g
Melting Point -3.0°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 202.0°C to 203.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CO2H
Refractive Index 1.4150 to 1.4180
Quantity 2.5 kg
Beilstein 02, 321
Merck Index 15, 1761
Specific Gravity 0.92
Solubility Information Solubility in water: 1.1% (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 3.23 mPa.s (20°C)
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Hexanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.