Learn More
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
22.11 € - 127.64 €
Chemical Identifiers
CAS | 142-61-0 |
---|---|
Molecular Formula | C6H11ClO |
Molecular Weight (g/mol) | 134.61 |
MDL Number | MFCD00000760 |
InChI Key | YWGHUJQYGPDNKT-UHFFFAOYSA-N |
Synonym | caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid |
PubChem CID | 67340 |
IUPAC Name | hexanoyl chloride |
SMILES | CCCCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10363432
|
Thermo Scientific Acros
169040050 |
5 g | Glass bottle |
22.11 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10723361
|
Thermo Scientific Acros
169041000 |
100 g | Glass bottle |
41.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10733361
|
Thermo Scientific Acros
169045000 |
500 g | Glass bottle |
127.64 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-61-0 | |
134.61 | |
YWGHUJQYGPDNKT-UHFFFAOYSA-N | |
67340 | |
CCCCCC(=O)Cl |
C6H11ClO | |
MFCD00000760 | |
caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid | |
hexanoyl chloride |
Specifications
142-61-0 | |
Colorless to Yellow | |
151.0°C to 153.0°C | |
Authentic | |
C6H11ClO | |
1.4253 to 1.4273 | |
02, 324 | |
0.95 | |
Solubility in water: hydrolysis. Other solubilities: decomposes in alcohol, soluble in ether, chloroform, ethylether and, acetone | |
CCCCCC(=O)Cl | |
134.61 | |
134.61 | |
Liquid |
-87.0°C | |
0.9500g/mL | |
53°C | |
96.5% min. (GC) | |
CH3(CH2)4COCl | |
MFCD00000760 | |
15, 1765 | |
caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid | |
YWGHUJQYGPDNKT-UHFFFAOYSA-N | |
hexanoyl chloride | |
67340 | |
97% | |
Hexanoyl chloride, 97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-549-1
missing translation for 'tsca' : TSCA
- Hexanoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.