Learn More
Hexyl isocyanate, 99%, Thermo Scientific Chemicals
46.90 € - 430.00 €
Chemical Identifiers
CAS | 2525-62-4 |
---|---|
Molecular Formula | C7H13NO |
Molecular Weight (g/mol) | 127.19 |
MDL Number | MFCD00037047 |
InChI Key | ANJPRQPHZGHVQB-UHFFFAOYSA-N |
Synonym | hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane |
PubChem CID | 75659 |
ChEBI | CHEBI:59058 |
IUPAC Name | 1-isocyanatohexane |
SMILES | CCCCCCN=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10405452
|
Thermo Scientific Acros
411660050 |
5 g | Glass Bottle |
46.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229890
|
Thermo Scientific Acros
411660250 |
25 g | Glass bottle |
150.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10117280
|
Thermo Scientific Acros
411661000 |
100 g | Glass bottle |
430.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2525-62-4 | |
127.19 | |
ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
75659 | |
1-isocyanatohexane |
C7H13NO | |
MFCD00037047 | |
hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
CHEBI:59058 | |
CCCCCCN=C=O |
Specifications
2525-62-4 | |
162°C to 164°C | |
Authentic | |
C7H13NO | |
1.4190 to 1.421 | |
04, IV, 718 | |
Solubility in water: decomposes | |
CCCCCCN=C=O | |
127.19 | |
CHEBI:59058 | |
99% | |
Hexyl isocyanate, 99% |
Colorless to Yellow | |
59°C | |
98.5% min. (GC) | |
CH3(CH2)5NCO | |
MFCD00037047 | |
hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
1-isocyanatohexane | |
75659 | |
127.19 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. May cause respiratory irritation. Causes serious eye damage. Harmful in contact with skin. Causes skin irritation. Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-763-8
missing translation for 'tsca' : TSCA
Recommended Storage : Product may develop some turbidity or precipitate
- Hexyl isocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only