missing translation for 'onlineSavingsMsg'
Learn More

Hexylamine, 99%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10368340
View Documents
Thermo Scientific Acros
204720050
5 mL Glass bottle N/A
N/A
10688504
View Documents
Thermo Scientific Acros
204721000
100 mL Glass bottle N/A
N/A
10318440
View Documents
Thermo Scientific Acros
204725000
500 mL Glass bottle N/A
N/A
10207123
View Documents
Thermo Scientific Acros
204720010
1 L Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-26-2
101.19
BMVXCPBXGZKUPN-UHFFFAOYSA-N
8102
hexan-1-amine
C6H15N
MFCD00008240
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
CHEBI:5712
CCCCCCN

Specifications

111-26-2
Colorless to Yellow
130.0°C
Authentic
C6H15N
1.4170 to 1.4190
04, 188
0.75
Solubility in water: 14g/L (20°C).
CCCCCCN
101.19
CHEBI:5712
101.19
Liquid
-19.0°C
0.7500g/mL
27°C
98.5% min. (GC)
CH3(CH2)5NH2
MFCD00008240
07,170
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
BMVXCPBXGZKUPN-UHFFFAOYSA-N
hexan-1-amine
8102
0.83 mPa.s (20°C)
99%
Hexylamine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-851-8

missing translation for 'rtecsNumber' : MQ4540000

missing translation for 'tsca' : TSCA

Product Identifier
  • Hexylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only