missing translation for 'onlineSavingsMsg'
Learn More
Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10714385

Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10714385
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10714385

Brand: Thermo Scientific Acros 427761000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3761-92-0, 109-99-9
Molecular Formula C6H13BrMg
Molecular Weight (g/mol) 189.38
MDL Number MFCD00000046
InChI Key SFXUWNMDTWQSHE-UHFFFAOYSA-M
Synonym hexylmagnesium bromide, magnesium, bromohexyl, n-hexylmagnesium bromide, n-hexmgbr, hexyl magnesium bromide, n-hexyl magnesium bromide, hexylmagnesium bromide solution, sfxuwnmdtwqshe-uhfffaoysa-m, hexylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagentShow MoreShow Less
PubChem CID 4184939
IUPAC Name magnesium;hexane;bromide
SMILES CCCCCC[Mg]Br

Specifications

Color Brown
Density 0.9800g/mL
Packaging AcroSeal™ Glass bottle
Linear Formula CH3(CH2)5MgBr
Quantity 100 mL
Specific Gravity 0.98
Formula Weight 189.39
Physical Form Liquid
Chemical Name or Material Hexylmagnesium bromide, 0.8M solution in THF

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466698 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number427761000
Lot NumberA0465800 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number427761000
Lot NumberA0448461 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number427761000,427768000
Lot NumberA0434708 Certificate TypeCertificate of Analysis Date11/10/2021 Catalog Number427761000,427768000
Lot NumberA0375672 Certificate TypeCertificate of Analysis Date02/02/2021 Catalog Number427761000,427768000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexylmagnesium bromide, 0.8M sloution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.