Learn More
Gibco™ Hibernate™-A Medium

Description
Hibernate-A Medium is a specialized CO2-independent nutrient medium designed for the maintenance and preservation of neural cells, tissues, and slices. Optimized for adult and postnatal neural samples, this medium supports neurons in ambient carbon dioxide conditions for up to 48 hours. This feature provides researchers with enhanced flexibility and control during experiments and transportation. Hibernate-A Medium allows you to run bench-top live cell experiments, such as microscopy, flow cytometry, and other physiology studies.
Benefits
- CO2 independence—eliminates the need for CO2 incubation for up to 48 hours, simplifying the maintenance and processing of neural cultures and tissues
- Convenience and flexibility—ideal for experiments that require extended handling times outside of CO2 incubators, offering greater convenience for researchers
- Transportation of neural samples—provides a reliable medium for the shipment of neural tissues, helping ensure their stability and viability upon arrival
Spécification
Spécification
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Form | Liquid |
| Product Type | Cell Culture Medium |
| Serum Level | Serum-free |
| Sterility | Sterile |
| With Additives | Phenol Red |
| Product Line | Hibernate |
| Quantity | 500 mL |
| Shipping Condition | Ambient |
| Afficher plus |
For Research Use Only. Not for use in diagnostic procedures.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.