missing translation for 'onlineSavingsMsg'
Learn More

HIF-1 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-449

535.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18090226

  • 658.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Canine, Chicken, Goat, Primate, Monkey samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIF-1 alpha
Polyclonal
Unconjugated
TBS, 0.1% BSA with 0.09% Sodium Azide
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Rabbit
93 kDa
0.1 mL
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
3091
Human, Mouse, Rat, Canine, Chicken, Goat, Primate, Monkey
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
0.2 mg/mL
Western Blot 1:2000 - 1:10000, Simple Western 1:200, Flow Cytometry 0.125 ug per 1 million cells in a 150 mcl reaction, ELISA 1:100-1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated, Knockdown Validated
Q16665
HIF1A
The immunogen recognized by this HIF-1 alpha Antibody maps to a region between residues 775 and the C-terminus (residue 826) of human hypoxia-inducible factor 1 [Uniprot# Q16665].
Affinity Purified
RUO
Primary
Monkey (COS-7) and Rat (review feedback). Has been shown to detect HIF-1 alpha in both mouse and human tissue by IHC with Citrate buffer antigen retrieval. Based on 100% sequence identity, this antibody is expected to react with: Cavia porcellus, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Grass carp, Northern pike, Atlantic cod, Duckbill platypus and Gansu zokor.
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only