Learn More
Novus Biologicals™ HIF-1 alpha Knockout Hypoxic-treated/untreated HeLa Cell Lysate
Cell Lysate
Brand: Novus Biologicals™
This item is not returnable.
View return policy
Specifications
HIF-1 alpha KO HeLa Whole cell lysate is provided as a Western blot positive control for Hif-1 research. The lysates are provided as a set of hypoxic and normoxic samples. Use 10 μL / lane for a standard mini-gel blot (approx. 1 mg/mL). | |
Human | |
Adaptive Immunity, Angiogenesis, Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Cytokine Research, HIF Target Genes, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1 A, HIF-1 alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 |
2 Vials | |
Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles. | |
HIF-1 alpha | |
HIF1A |