Learn More
Applied Biosystems™ High-Capacity RNA-to-cDNA™ Kit
The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes.
308.00 € - 2790.00 €
Specifications
Format | Kit |
---|---|
Reaction Speed | 30 to 60 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 37°C |
Reverse Transcriptase | MultiScribe |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10704217
|
Applied Biosystems™
4387406 |
50 Reactions |
308.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16271891
|
Applied Biosystems™
4388950 |
500 Reactions |
2790.00 €
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes. The components of the two-tube kit work together to provide sensitive and specific reverse transcription across a broad range of template amounts.
Features of the High Capacity RNA-to-cDNA Kit include:
• Fast reaction time (typically 30–60 min)
• Convenient and simple workflow with few pipetting steps
• Reliable reverse transcription of both abundant and limited targets
• Optimized two-step protocol enables multiple PCR reactions from a single RT reaction
Streamline cDNA synthesis while maintaining detection sensitivity
Amplification of a dilution series of the RNA concentration reference standard demonstrates exceptional reverse transcription and PCR efficiency across 11 orders of magnitude. This wide dynamic range allows one set of reaction conditions to detect transcripts from highly active as well as weakly expressed genes. The two-tube formulation reduces reaction time and requires fewer pipetting steps than with the High Capacity cDNA Reverse Transcription Kit, while maintaining high performance.
Specifications
Kit | |
Reverse Transcription | |
MultiScribe | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
30 to 60 min. | |
37°C | |
Yes | |
Real Time PCR (qPCR), RT-PCR | |
Premixed Components | |
Up to 7 kb |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.