Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Resin
Strep-Tactin™ Superflow™ High Capacity Resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
191.00 € - 28550.00 €
Specifications
Product Type High Capacity Resin Suspension
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12856456
View Documents
IBA Lifesciences
2-1208-002
4 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12866456
View Documents
IBA Lifesciences
2-1208-010
20 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12876456
View Documents
IBA Lifesciences
2-1208-025
50 mL
1610.00 €
50mL
In Stock
to see stock.
Edge
Add to basket
12896456
View Documents
IBA Lifesciences
2-1208-500
1000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Strep-Tactin™ Superflow™ high capacity is a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.  
Specifications
Specifications
High Capacity Resin Suspension