missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole Parmer High-Capacity Vacuum Pump, Gauge/Reg
Small, quiet source of vacuum or pressure
Brand: Cole Parmer 07061-42
745.44 EUR valid until 2024-12-31
Use promo code "21962" to get your promotional price.
This item is not returnable.
View return policy
Description
Small, quiet source of vacuum or pressure
- Free-air capacities up to 2.2 cfm (62.3 L/min)
- Shaded-pole, fan-cooled motor has thermal overload switch to protect your pump system
- Different pump models have heads in parallel for higher capacity or series for greater vacuum
- Select vacuum pumps with or without gauges, regulators, and valves
- Runs dry
Specifications
24 Hg | |
High Capacity Vacuum Pump | |
60 psi |
28.3 L/min. | |
Gast line of oil-less diaphragm air compressors and vacuum pumps | |
220 V |