Learn More
Contec™ High Duster XF with Hand Grip
The High Duster XF makes cleaning hard-to-reach spaces easier when paired with MicroGenesis Microfiber High Duster Covers. The flexible body of the high duster tool bends to hug surface contours and reduces operator strain.
Brand:  Contec™ MGMP0008
Product Code. 30274784
  • 90.09 € / Each
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flexible frame reaches areas where typical dusters cannot
  • Reduces cross contamination risk when paired with a Microfiber High Duster Cover
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Duster Tool
No
24.25 in.
62 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Contec™ High Duster XF with Hand Grip > High Duster XF; Flammable: No; Product Type: Duster Tool; Length (English): 24.25 in.; Length (Metric): 62 cm