missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente High-Efficient Condensers with Four Coils
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1715.00 € - 1785.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10284481
|
Lenz Laborglasintrumente
05255000 |
Cone and socket NS 45/50 |
1715.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||||
11596814
|
Lenz Laborglasintrumente
05255050 |
Ball and socket KF 50 |
1785.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
- High-efficient condensers with four cooling coils
- Cooling area: 2,350 cm²
- Jacket length: 300 mm
- Made of DURAN™ tubing
- Packaging unit: 1 piece
Specifications
Duran Glass | |
300 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title