Learn More
Thermo Scientific Chemicals High Form Nickel Crucible
Brand:  Thermo Scientific Chemicals 035883.KT
33.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15493575
  • 34.07 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High Form
20 mL
35mm
22mm
Nickel
38mm
Nickel Crucible