Learn More
Heidolph Instruments High Hood XL
For use as an add on to incubating shakers
Brand:  Heidolph Instruments 549-90060-00
Product Code. 10792014
  • 1515.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With a height of 428 mm, this cap is particularly suitable for tall culture vessels, such as 2000 mL Erlenmeyer flasks
Specifications
Specifications
115/230 V
5°C to 65°C
428 mm