missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ High Impact Polystyrene Inoculating Loop

Product Code. p-8010214
Click to view available options
Volume:
1 μL
5 μL
10 μL
Color:
Red
White
Yellow
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12759865

Brand: Technical Service Consultants™ TS/1A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Technical Service Consultants™ High Impact Polystyrene Inoculating Loop for general laboratory needs including plating and quick and accurate sample throughput.

  • Cadmium-free pigments
  • Gamma Irradiation to achieve a SAL of 10-6
  • Shelf Life: 3 years
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container

Specifications

Disposable Single-use
Volume 10 μL
Material High-impact Polystyrene
Sterility Sterile
Color White
Product Type Inoculating Loop
Quantity 1000/Pk.
Packaging Type Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.