missing translation for 'onlineSavingsMsg'
Learn More

Merck High Output Pumps

Features a piston-driven design for greater power

Brand:  Merck WP6222050

 View more versions of this product

Product Code. 10542975

  • 2945.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Features a piston-driven design for greater power

Supplied with 70cm of 1/4 in. tubing and a Millex FA50 filter for in-line moisture protection

  • Cast Aluminum construction
  • 1/4 in. Stepped hose barb
  • Filtration Supply Type: Vacuum Pump

Certifications

UL listed, CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

70cm of 0.25 in. dia. tubing, Millex™ FA50 filter for in-line moisture protection
220 V 50 Hz
11.7 lb.
34 L/min.
8 in.
10 in.
9 in.
5.4 bar
UL, CE
Cast Aluminum
5.3 kg
50 Hz
20.3 cm
25.4 cm
High Vacuum Pump
220 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.