Learn More
Sartorius™ High Performance Glass Shield Secura26
High performance glass shield for use with laboratory balance.
Brand:  Sartorius™ YHK01SQP
Product Code. 17711828
  • 984.00 € / Each
Explore more special offers
Description
Description
  • Used for increasing weighing performance of balances with readability of 0.002 mg
  • (Supplied without a balance)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Secura Laboratory Balances
Draft Shield