Learn More
Cole-Parmer™ High-Performance Ultraviolet Viewing Cabinet
Use this viewing cabinet for optimal sample viewing with uniform UV illumination. Cole-Parmer™ High-Performance Ultraviolet Viewing Cabinets feature high-intensity 15-watt UV tubes. Control switches for selecting UV or white light are conveniently located on the top of the cabinet.
Brand:  Cole-Parmer™ 97621-25
Product Code. 15415104
  • 3395.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Intensity: 2000 μW/cm2
  • Soft rubber viewport provides a comfortable viewing area with adequate space for eyeglasses
  • Block out ambient light with lightweight access curtain
  • Protect your eyes with contrast control filter 
  • Removable bottom panel allows cabinet to be placed over large samples or a transilluminator
  • Lightweight access curtain blocks out ambient light
  • Model features four longwave UV tubes mounted on one side of the cabinet
Specifications
Specifications
UV Viewing Cabinet
365 nm
4 x 15 W, 230 V, 50 Hz
4 x 15 W
23.375 x 15.75 x 13 in.