Learn More
Brannan™ Precision Lab Thermometer with 3-points Traceable Calibration
High precision lab thermometer complete with calibration certificate.
Brand:  Brannan™ 50/521/0
 View more versions of this product
Product Code. 17863298
  • 110.00 € / Each
Explore more special offers
Description
Description
  • A range of spirit filled, high precision thermometers which are supplied with a calibration certificate
  • Safe non-mercury filling
  • Presented in a study cardboard cylinder for storage
  • Made in UK
Specifications
Specifications
0°C, 35°C, 90°C
Partial Immersion
Glass
230°F
-4°F
-4°F to +230°F
± 1.0°C
76 mm
UK Traceable to National Standards ISO 9001
30.5 cm
Blue Spirit
110°C
-20°C
-20°C to +110°C
0.5°C
1.2 cm