Learn More
3M™ Petrifilm™ High-Sensitivity Coliform Count Plate
Accommodates 5 mL samples and detection of low levels of confirmed coliforms in 24 hours.
132.80 € - 1193.43 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17223754
|
3M™ Petrifilm™
7100039357 |
50 |
132.80 €
Pack of 50 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
17253754
|
3M™ Petrifilm™
7100039398 |
500 |
1193.43 €
Pack of 500 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
Description
- Coliform results in 24 hours.
- Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
- Unique plate design to trap the gas produce by coliform
- This plate has been awarded Official Methods of Analysis (OMA) #996.02 for Dairy products and NF VALIDATION certificate: 3M-01/7-03/99 for Human food products by AFNOR Certification.
- Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Food and Beverage, Environmental Test |