Learn More
Fisherbrand™ High Speed Module
High speed module consist of 2 tanks on top of the machine.
Specifications
Product Type High Speed Module
Includes 2 Tanks
View More Specs
Description
Description
  • Tanks electrically heated
  • Water is preheated before going into the washing chamber, allowing for shorter cycle times
Specifications
Specifications
High Speed Module
2 Tanks