missing translation for 'onlineSavingsMsg'
Learn More

Scotch™ High Tack Box Sealing Tape 375+

This high-performance packaging tape securely closes a variety of heavy weight box materials, including recycled fiberboard. The polypropylene film backing is easy to unwind and resists abrasion, moisture, scuffing and edge tear.

Brand:  Scotch™ 7100215092

45.01 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 17701679

  • 50.00 € / 6 rolls
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-performance hot melt packaging tape for heavy duty applications
  • Strong adhesive helps prevent tampering and tape failure during shipping and delivery
  • High edge tear and split resistance for e-commerce and single-parcel shipments
  • Synthetic hot melt rubber adhesive with polypropylene backing for conformability
  • Easy unwind and smooth dispensing for use on high-speed automated packaging lines
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hot-melt
Tape
Box Sealing Tape
36 Pk.
164 ft.
1.88 in.
Transparent
Polypropylene
Heavy Duty Packaging
50 m x 48 mm
50 m
48 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.