Learn More
Brady™ High Temp, Flame Retardant Polyimide Wrap Around Wire and Cable Label Tape for M7 Printers
Flame Retardant Wire Wraps are ideal for cable and wire bundle applications and label applications where self-extinguishing properties are required.
Brand:  Brady™ 173261
 View more versions of this product
Product Code. 17829134
  • 343.00 € / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyimide material specifically designed to self-extinguish and prevent flames from propagating
  • Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed
  • Service temperature of -40°F to 212°F (-40°C to 100°C)
  • Made of B-472 polyimide material
Specifications
Specifications
Not Autoclavable
M7C-2000-472-YL
None
Matte
Polyimide
Thermal Transfer
Non-sterile
M710, BMP71
50 ft.
-94°F to +212°F
0.0032 in.
50 ft. x 2 in.
15.24 m
50.8 mm
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AS-81531
Permanent Acrylic
Yellow
Continuous
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Wire and Cable Labeling
No
Rectangle
-70°C to +100°C
0.081 mm
50 ft.
2 in.