missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ High Temperature Furnace LHT/D Series

Compact tabletop models are perfectly suited for the sintering of technical ceramics, such as zirconium oxide dental bridges.

Brand:  Nabertherm™ LHT327VN1

 View more versions of this product

Product Code. 17327989

  • 11010.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Maximum Temperature: 1650°C
  • High-quality molybdenum disilicide heating elements
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Compact design with lift door, opening upwards
  • Adjustable air inlet
  • Exhaust air opening in the roof
  • Type B thermocouple
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications

Specifications

P570
1650°C
630 x 470 x 1020 mm
3 L
155 x 135 x 200 mm
60 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers