Learn More
High Vacuum Suction Machine Sam14 Static Unit
Brand:  N/A SAM14/01010102
Encompass
Product Code. 12937934
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Minor operating theatre with 2 x 2L autoclavable cannister 220-240V 50/60Hz 650mm x 200mm x 380mm (w x d x h) 30L/min flow rate, 0 to 650mmHg vacuum