Learn More
High Vacuum Suction Machine Sam36 Mobile Unit
Brand:  N/A SAM36/01010102
Encompass
Product Code. 12997934
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Inter-uterine suction with 2 x 2L autoclavable cannister 220-240V 50/60Hz 390mm x 330mm x 850mm (w x d x h) 25L/min flow rate, 0 to 730mmHg vacuum