Learn More
Manutan High visibility cargo trouser, 40 regular, yellow
Product Code. 12829679
Encompass_Preferred
Water and stain repellent finish with triple stitched inner and outer leg seems for added strength. Internal knee pad pockets. Metal button fastener with YKK Brass zip fly. Side waist elastication and five belt loops for a comfortable fit. 280g poly/cotton fabric, the cotton allows the fabric to breathe, while the polyester makes it durable and easy to care for. EN/ISO 20471 Class 1, low level of visibility when worn without other high visibility clothing.
  • 40 waist size
  • Yellow
  • Length: regular L (31in)