Learn More
Manutan High visibility waterproof anorak, yellow, medium
Product Code. 12854269
Encompass_Preferred
Yellow waterproof anorak with a warm quilt lining, integral hood and a shaped drop back. Full length Velcro fastened storm flap, keeps your dry in wind driven rain. Two-way main zip, for added convenience and comfort. Hood in collar and lycra storm cuff. Velcro fastened front pockets, mobile phone pocket and shaped drop back. PU coated polyester. Complies with Railway Group Standard GO/RT 3279. EN 343 Level 3, greatest protection against rain. EN 343 Level 1, low level of breathability. EN 20471 Class 3, highest protection level providing maximum visibility.
  • Medium
  • Chest size: 36-39
  • Yellow