Learn More
Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand:  Hamilton™ 235904_CORE_I
Product Code. 10666025
  • 327.00 € / Pack of 3840
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 95.1mm tip length
  • Color code label: white
  • Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
  • Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
Specifications
Black
1000 μL
Non-sterile