missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ 26/600 Superdex™ 200 pg

Use for separating MAbs from dimers and contaminants. Cytiva Life Sciences™ HiLoad™ 26/600 Superdex™ 200 pg is ideal for high-resolution preparative gel filtration chromatography.

Brand:  Cytiva 28-9893-36

3760.58 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



Product Code. 11390342

  • 4231.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes:
  • Dismantling tool
  • Support screen
  • Net ring (10μm)
  • O-ring
  • Stop plugs
  • Instructions
This item is not returnable. View return policy

Description

Description

Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose. Dextran component determined the separation properties of these media. Steep selectivity curves give gppd resolving power for peptides and proteins in the molecular weight range Mr 10 000 600 000 (Superdex 200 pg).

  • Used for separating MAbs from dimers and contaminants
  • Short run times with high recovery
  • Medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions
  • Separations can be scaled up to production levels
  • 0.06 in. fittings to connect to AKTA design systems
  • Theoretical plates: >13000m-1
  • Flow rate: <4.4mL/min.
  • Flow velocity:     10m/h to 50cm/h1
  • Pressure max. (over the packed bed during operation): 3 bar [0.3MPa] (42psi)

  • Media
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH Stability working range: 3 to 12
  • pH Stability cleaning range: 2 to 14
  • Fractionation [Mr] globular proteins: 1 × 104-6 × 105
  • Fractionation [Mr] dextrans: 1 × 103-1 × 105
  • Shelf life from manufacture date: 5 years
  • Chemical stability: all commonly used buffers, 1M NaOH, 0.1M HCl, 1M acetic acid, 8M urea, 6M guanidine HCl, 1% SDS, 70% ethanol, 30% iso-propanol, 30% acetonitrile
  • Column
  • Column I.D.: 1.02 in. (26mm)
  • Inlet I.D.: 0.031 in. (0.8mm)
  • Outlet I.D.: 0.031 in. (0.8mm)
  • Chemical stability: resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol
  • Column hardware made from borosilicate glass, polyamide, fuororubber, Ethylene Propylene Diene (EPDM), ETFE, Superpolyoxymethylene
  • Column tube made from borosilicate glass
  • Bed support made from Polypropylene (PP) (reinforced glass fiber), Polyamide (PA)
  • Porosity: 10μm

Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids

Order Info

HiLoad columns are prepacked XK columns; Bed volume: 320mL; Bed dimensions: 1.02 x 23.62 in. (26 x 600mm); Sample volume: 13mL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs, and instructions.
13 mL
4°C to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiLoad 26/600 Superdex 200 pg

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.