missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ Superdex™ 200 PG XK Column

Prepacked XK columns designed for high-resolution preparative gel filtration chromatography.

Brand:  Cytiva 90-1001-37


 View more versions of this product

Product Code. 11389991

  • 5473.00 € / Each
Estimated Shipment: 10-10-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

HiLoad™ Superdex™ 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.

  • High resolution with short run times and good recovery.
  • High capacity at high flow rates.
  • Separations can be scaled up to production levels.
  • Prepacked for convenience and reproducibility.
  • 1/16” fittings for convenient connections to ÄKTA design systems.

Superdex™ prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these resins are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex™ 30 pg), Mr 3 000 to 70 000 (Superdex™ 75 pg), and Mr 10 000 600 000 (Superdex™ 200 pg). The resin combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Pk.
24 to 44 μm
Superdex Pre-packed Column
900 mm
3bar
Composite of Cross-Linked Agarose and Dextran
4 to 30°C, 20% Ethanol
10 to 50 cm/hr.
180mL
16 mm
Borosilicate Glass
3 to 12
Dismantling Tool, Support Screen, Net Ring (10μm), o-Ring, Stop Plugs and Instructions
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Superdex 200 prep grade, XK 16/100, 90-95 cm

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.