missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiPrep™ 26/10 Desalting Columns
Provide reliable and reproducible desalting and buffer exchange with sample sizes of up to 15mL per column. Cytiva HiPrep 26/10 Desalting Column is a convenient, easy-to-use column prepacked 53mL column with Sephadex G-25 Fine for desalting and buffer exchange.
725.00 € - 2486.00 €
Specifications
Description | HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange. |
---|---|
Product Type | HiPrep 26/10 Desalting |
For Use With (Equipment) | Laboratory pump or chromatography system such as AKTA design |
Particle Size | >20 μm (dry), 17 to 132 μm (wet) |
Diameter (English) Inner Column | 1.02 in. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10470505
|
Cytiva
17-5087-02 |
4 x 53 mL |
2486.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10460505
|
Cytiva
17-5087-01 |
1 x 53 mL |
725.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Saves time and labor as replacement for dialysis
- Two to four columns can be used in series for sample sizes 30 to 60mL
- Compatible with a laboratory pump or a chromatography system
- Fittings are 1/16 in. for easy connection to AKTAdesign systems
- Long-term and working pH range 2 to 13
- Maximum flowrate: 15mL/min.
Specifications
HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange. | |
Laboratory pump or chromatography system such as AKTA design | |
1.02 in. | |
4°C to 30°C in 20% Ethanol | |
Reusable |
HiPrep 26/10 Desalting | |
>20 μm (dry), 17 to 132 μm (wet) | |
53 mL | |
26 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiPrep 26/10 Desalting, 4 pack
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16,5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HiPrep™ 26/10 Desalting Columns