missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ HIC FF 16/10 Columns

Produce fast, preparative separation of proteins and other biomolecules. Cytiva Life Sciences™ HiPrep™ HIC FF 16/10 Columns are high capacity with excellent flow properties, and reliable, reproducible separations.

508.00 € - 759.00 €

Specifications

Bed Volume 20mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20 mL
Height (English) 3.93 in.
View More Specs

Products 4
Product Code Brand Product Type Support Type Price Quantity & Availability  
Product Code Brand Product Type Support Type Price Quantity & Availability  
10545905
View Documents
Cytiva
28-9365-48
Octyl Sepharose 4% Cross-linked Agarose
759.00 €
Each
In Stock
to see stock.
Add to basket
 
10792227
View Documents
Cytiva
28936546
Phenyl Sepharose (low sub) 6% Cross-linked Agarose This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10608225
View Documents
Cytiva
28-9365-45
Phenyl Sepharose (high sub) 6% Cross-linked Agarose
759.00 €
Each
In Stock
to see stock.
Add to basket
 
10054334
View Documents
Cytiva
28-9365-47
Butyl Sepharose 4% Cross-linked Agarose
759.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

The 20mL HIC columns provide fast, preparative separation of proteins and other biomolecules. They are ready-to-use with four media choices: Phenyl Sepharose 6 Fast Flow (high sub); Phenyl Sepharose 6 Fast Flow (low sub); Butyl Sepharose 4 Fast Flow; and Octyl Sepharose 4 Fast Flow.

Features:

  • Offer high capacity, excellent flow properties and reliable, reproducible separations
  • Compatible with simple pump-based configurations or chromatography systems (e.g., AKTA)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol
  • Storage temperature: 4° to 30°C

Specifications

Specifications

20mL
16mm
3.93 in.
>300 cm/hr.
5 bar
45 to 165μm
Ion Exchange Column
0.62 in.
1 x 20 mL
100mm
Polypropylene
3 to 13
4° to 30°C
Provide fast, preparative separation of Proteins and other biomolecules by hydrophobic interaction chromato graphy (HIC), initial and intermediate purification, removing bulk impurities
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep Octyl FF 16/10

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.