missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ IEX FF 16/10 Columns

Provide fast, reliable and reproducible preparative ion exchange separations

733.00 €

Specifications

Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Bed Volume 20mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20mL
View More Specs

Products 2
Product Code Brand Ligand Type Max. Pressure pH Range Price Quantity & Availability  
Product Code Brand Ligand Type Max. Pressure pH Range Price Quantity & Availability  
10607855
View Documents
Cytiva
28-9365-41
Diethylaminoethyl 5 bar 2 to 12
733.00 €
Each
In Stock
to see stock.
Add to basket
 
10617855
View Documents
Cytiva
28-9365-42
Carboxymethyl 1.5 bar 6 to 10
733.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

DEAE and CM Sepharose™ 6 Fast Flow are prepacked in 20mL HiPrep columns. Ideal for rapid enrichments during initial capture of proteins from starting material.

Features:

  • Combine good flowrates and high binding capacity
  • Simple-to-operate
  • Compatible with single-pump configurations and other chromatography systems (e.g., AKTA™)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol (DEAE,CM), 0.2M sodium acetate in 20% ethanol (SP)
  • Storage temperature: 4° to 30°C

Specifications

Specifications

1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
0.62 in.
1 x 20mL
100mm
Polypropylene
6% Cross-linked Agarose
Ion Exchange Column
20mL
16mm
3.93 in.
>300 cm/hr.
45 to 165μm
4° to 30°C
Preparative separations of Proteins and other biomolecules using ion exchange (IEX) chromato graphy
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep DEAE FF 16/10

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.