missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiPrep™ Sephacryl™ S-100 HR Columns
Maximum recovery and minimum nonspecific adsorption
865.00 € - 1248.00 €
Specifications
Quantity | Each |
---|---|
Particle Size | 25 to 75 μm |
For Use With (Equipment) | Low to medium pressure AKTA™ systems |
Product Type | Sephacryl HR Column |
For Use With (Application) | Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix |
Product Code | Brand | Flow Rate | Bed Volume | Diameter (Metric) Inner | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Bed Volume | Diameter (Metric) Inner | Price | Quantity & Availability | |||
10369334
|
Cytiva
17-1194-01 |
<1.3 mL/min. | 320 mL | 2.6cm |
1248.00 €
Each |
In Stock
Log in to see stock. |
|||
10146364
|
Cytiva
17-1165-01 |
<0.5 mL/min. | 120 mL | 1.6cm |
865.00 €
Each |
In Stock
Log in to see stock. |
|||
Description
- Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Recommended linear flowrate 5 to 15cm/hr.
Designed for separating peptides and small proteins
Specifications
Each | |
Low to medium pressure AKTA™ systems | |
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix | |
60cm | |
1.5 bar [0.15 MPa] (22 psi) | |
Allyl Dextran and N, N'-methylenebisacrylamide | |
4°C to 30°C, 20% Ethanol |
25 to 75 μm | |
Sephacryl HR Column | |
23.62 in. | |
Polypropylene | |
3 to 11 | |
Peptides and small proteins |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiPrep 26/60 Sephacryl S-100 HR
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.