missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Sephacryl™ S-200 HR Columns

Maximum recovery and minimum nonspecific adsorption

Brand:  Cytiva 17-1166-01

771.01 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 10106414

  • 865.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.

Designed for purifying antibodies, serum proteins and midsize proteins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each
25 to 75 μm
120 mL
1.6cm
23.62 in.
Polypropylene
3 to 11
Antibodies, serum proteins and mid-size proteins
<0.5 mL/min.
Low to medium pressure AKTA™ systems
Sephacryl HR Column
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
60cm
1.5 bar [0.15 MPa] (22 psi)
Allyl Dextran and N, N'-methylenebisacrylamide
4°C to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep 16/60 Sephacryl S-200 HR

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.