Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
260.00 € - 2257.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11540894
|
Cytiva
28-9673-90 |
5 x 1 mL |
260.00 €
Pack of 5 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
11545804
|
Cytiva
28-9799-17 |
10 x 1 mL | N/A | ||||||
17296876
|
Cytiva
29104065 |
100 mL |
2257.00 €
100mL |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
Description
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
- High-capacity small-scale purification of histidine-tagged proteins from different sources
- High purity and yield
- Easy parallel screening of histidine-tagged proteins with high repeatability
- Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
Order Info
1mL of medium slurry is sufficient for five purification runs according to the recommended protocol
Specifications
50 mg Histidine-tagged protein/mL medium |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Flammable liquid Category 3