Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
Brand:  Cytiva 28-9673-88
Product Code. 11530894
  • 110.53 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
  • High-capacity small-scale purification of histidine-tagged proteins from different sources
  • High purity and yield
  • Easy parallel screening of histidine-tagged proteins with high repeatability
  • Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mg Histidine-tagged protein/mL medium
2 x 1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3