missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ Q ImpRes and Capto™ SP ImpRes

Product Code. p-4592283 Shop All Cytiva Products
Click to view available options
Product Type:
Capto Q ImpRes medium
Capto SP ImpRes medium
Ligand Type:
Quaternary Amine
Sulphonate group
pH Range:
2 to 12
4 to 12
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11310002

Brand: Cytiva 17546802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands

  • Delivers pressure-flow properties with wide resolution
  • High-throughput purifications easy to optimize and scale up
  • High flow agarose based matrix
  • Runs at high flow rates as well as high bed heights thus increasing flexibility in process design
  • Developed for large-scale purification of biopharmaceutical proteins
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Available in a wide range of different bulk pack sizes and pre-packed formats for process development

Capto Q ImpRes: Strong anion (quaternary amine group) exchange BioProcess™ medium
Capto SP ImpRes: Strong cation (sulfonate group) exchange BioProcess medium

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type Capto SP ImpRes medium
Diameter (English) Inner 39.3 in.
Diameter (Metric) Inner 1000mm
Quantity 100 mL
Height (English) 7.87 in.
Height (Metric) 200mm
Flow Rate 220 cm/hr.
Ligand Type Sulphonate group
Max. Pressure <3 bar
pH Range 4 to 12
Particle Size 36 to 44μm
Support Type High-flow Agarose
Content And Storage 4° to 30°C
For Use With (Application) High-throughput intermediate purification and polishing steps of a wide range of biomolecules
Matrix Capto Q Chromatography Media
Show More Show Less
compliance-icons
Product Identifier
  • Capto SP ImpRes, 100 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.