missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns

Designed for method optimization and parameter screening for capture of monoclonal antibodies. Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns come ready-to-use for convenience.

Brand:  Cytiva 28-9269-73


 View more versions of this product

Product Code. 10811291

  • 1037.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ready-to-use columns for convenient process development
  • Rigid, highly cross-linked agarose matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

MabSelect

  • For capture of MAbs from large sample volumes
  • Prepacked with protein A-derived media
  • Compatible with high flow rates and high pressure when scaling up
MabSelect Xtra
  • Increased binding capacity enables purification of MAbs from high-level expression feedstock
  • Also optimized for Fc-fusion proteins
  • Very low, unspecific binding due to high ligand selectivity and matrix hydrophilicity
  • Prepacked with protein A-derived media
MabSelect SuRe (Superior Resistance) and MabSelect SuRe (Superior Resistance) LX
  • With novel, alkali-tolerant recombinant protein A ligand that provides greater stability and allows use of 0.1 to 0.5M sodium hydroxide for cleaning-in-place (CIP)
  • Enhanced protease resistance resulting in lower ligand leakage
  • High dynamic binding capacity

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol
Bed Dimensions: 7.7 × 100 mm
1 x 4.7mL
0.3 in.
0.77cm
3.93 in.
Antibody affinity chromatography
Epoxy Activation
85μm
3 to 10
4.7 mL
HiScreen MabSelect Prepacked Column
Stoppers at the inlet and outlet
Filter
≈30mg Human IgG
HiScreen MabSelect Prepacked Column
100 mm
Recombinant Protein A (E.coli)
Polypropylene
3 bar [0.3 MPa] (44 psi)
2° to 8°C
Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen MabSelect

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.